Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubqln3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107543.2 | NP_001101013.1 | 1 - 2 | 63 | 72 | |
GenBank mRNA | BC161885.1 | - | 1 - 2 | 63 | 7133921 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons |