Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekhb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006229787.2 | XP_006229849.1 | 2 - 3 | 381 | 125 | |
XM_006229788.2 | XP_006229850.1 | 1 - 2 | 542 | 125 | ||
XM_006229789.2 | XP_006229851.1 | 2 - 3 | 452 | 125 | ||
XM_008759795.1 | XP_008758017.1 | 1 - 2 | 538 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |