Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekhb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006229791.2 | XP_006229853.1 | 5 - 6 | 1390 | 72 | |
XM_008759795.1 | XP_008758017.1 | 5 - 6 | 914 | 72 | ||
GenBank mRNA | BC078704.1 | - | 5 - 6 | 549 | 7124581 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Rattus norvegicus Probe spans exons |