Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gdpd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006229796.2 | XP_006229858.1 | 6 - 7 | 643 | 67 | |
XM_006229799.2 | XP_006229861.1 | 6 - 7 | 641 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |