Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077675.1 | NP_001071143.1 | 6 - 7 | 945 | 111 | |
GenBank mRNA | BC127539.1 | - | 6 - 7 | 945 | 7192024 | 111 |
FQ224850.1 | - | 6 - 7 | 986 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons |