Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Calca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033955.1 | NP_001029127.1 | 4 - 5 | 535 | 121 | |
NM_001033956.1 | NP_001029128.1 | 4 - 5 | 511 | 121 | ||
GenBank mRNA | AW523601.1 | - | 1 - 2 | 121 | ||
AY702025.1 | - | 1 - 2 | 171 | 121 | ||
CB746609.1 | - | 2 - 3 | 197 | 121 | ||
CF978038.1 | - | 4 - 5 | 517 | 121 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |