Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bysl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182674.3 | NP_872615.3 | 3 - 4 | 634 | 101 | |
GenBank mRNA | BC079030.1 | - | 3 - 4 | 623 | 7114086 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |