Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rrp36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002729998.4 | XP_002730044.2 | 4 - 5 | 595 | 120 | |
XM_006244572.2 | XP_006244634.1 | 4 - 5 | 598 | 120 | ||
XM_006244573.2 | XP_006244635.1 | 5 - 6 | 460 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |