Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024360.2 | NP_001019531.1 | 1 - 2 | 367 | 66 | |
XM_008766878.1 | XP_008765100.1 | 1 - 2 | 1466 | 66 | ||
GenBank mRNA | AB210855.1 | - | 1 - 2 | 267 | 66 | |
BC128712.1 | - | 1 - 2 | 367 | 7133504 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |