Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crisp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039393.2 | NP_001034482.2 | 6 - 7 | 702 | 82 | |
XM_008766935.1 | XP_008765157.1 | 6 - 7 | 702 | 82 | ||
GenBank mRNA | DQ200907.1 | - | 6 - 7 | 702 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Protease Rattus norvegicus Probe spans exons |