Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfap2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106896.2 | NP_001100366.2 | 1 - 2 | 331 | 86 | |
XM_006244649.1 | XP_006244711.1 | 1 - 2 | 331 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |