Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps18a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198756.2 | NP_942051.1 | 2 - 3 | 149 | 153 | |
GenBank mRNA | BC058507.1 | - | 2 - 3 | 153 | 6889538 | 153 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |