Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109303.1 | NP_001102773.1 | 7 - 8 | 567 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |