Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnq5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134643.2 | NP_001128115.2 | 1 - 2 | 573 | 90 | |
GenBank mRNA | BE103175.1 | - | 249 | 90 | ||
FQ084615.1 | - | 1 - 2 | 214 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Ion Channel Rattus norvegicus Probe spans exons |