Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rsph9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006244538.2 | XP_006244600.1 | 4 - 5 | 1248 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Rattus norvegicus Probe spans exons |