Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rcan2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006244593.2 | XP_006244655.1 | 1 - 2 | 374 | 69 | |
GenBank mRNA | AF459022.1 | - | 1 - 2 | 172 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |