Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptcra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003750652.3 | XP_003750700.1 | 3 - 4 | 1031 | 116 | |
XM_008766898.1 | XP_008765120.1 | 3 - 4 | 678 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |