Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ckmt1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012738.1 | NP_001012756.1 | 6 - 7 | 1214 | 63 | |
XM_006234830.2 | XP_006234892.1 | 7 - 8 | 1170 | 63 | ||
GenBank mRNA | BC091335.1 | - | 6 - 7 | 1214 | 7314893 | 63 |
X59737.1 | - | 7 - 8 | 1124 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |