Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspa12b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107778.1 | NP_001101248.1 | 2 - 3 | 183 | 111 | |
XM_006235027.2 | XP_006235089.1 | 3 - 4 | 338 | 111 | ||
XM_006235028.2 | XP_006235090.1 | 2 - 3 | 218 | 111 | ||
XM_008762178.1 | XP_008760400.1 | 2 - 3 | 1259 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |