Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013919.1 | NP_001013941.1 | 3 - 4 | 343 | 81 | |
XM_006234904.2 | XP_006234966.1 | 2 - 3 | 798 | 81 | ||
XM_006234906.2 | XP_006234968.1 | 2 - 3 | 303 | 81 | ||
GenBank mRNA | BC083716.1 | - | 3 - 4 | 343 | 7190694 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |