Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tank | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164073.1 | NP_001157545.1 | 4 - 5 | 454 | 129 | |
NM_145788.2 | NP_665731.2 | 4 - 5 | 465 | 129 | ||
GenBank mRNA | AJ422211.1 | - | 3 - 4 | 325 | 129 | |
BC129086.1 | - | 4 - 5 | 454 | 7365827 | 129 | |
EV776506.1 | - | 4 - 5 | 465 | 129 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |