Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zswim2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011960.1 | NP_001011960.1 | 6 - 7 | 866 | 117 | |
XM_006234435.2 | XP_006234497.1 | 6 - 7 | 900 | 117 | ||
GenBank mRNA | BC079431.1 | - | 6 - 7 | 866 | 7114765 | 117 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |