Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108956.1 | NP_001102426.1 | 5 - 6 | 583 | 67 | |
XM_008762068.1 | XP_008760290.1 | 5 - 6 | 529 | 67 | ||
XM_008762069.1 | XP_008760291.1 | 5 - 6 | 526 | 67 | ||
XM_008762070.1 | XP_008760292.1 | 5 - 6 | 1875 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |