Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifih1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109199.1 | NP_001102669.1 | 12 - 13 | 2681 | 64 | |
GenBank mRNA | BC168680.1 | - | 9 - 10 | 1706 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons |