Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcna | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022381.3 | NP_071776.1 | 5 - 6 | 767 | 94 | |
GenBank mRNA | BC060570.1 | - | 5 - 6 | 852 | 6891516 | 94 |
FQ219897.1 | - | 5 - 6 | 848 | 94 | ||
FQ220101.1 | - | 5 - 6 | 848 | 94 | ||
FQ220309.1 | - | 5 - 6 | 848 | 94 | ||
FQ220835.1 | - | 5 - 6 | 848 | 94 | ||
FQ220924.1 | - | 5 - 6 | 847 | 94 | ||
FQ225495.1 | - | 5 - 6 | 848 | 94 | ||
FQ225620.1 | - | 4 - 5 | 472 | 94 | ||
FQ226677.1 | - | 5 - 6 | 847 | 94 | ||
FQ226726.1 | - | 5 - 6 | 849 | 94 | ||
FQ227521.1 | - | 5 - 6 | 848 | 94 | ||
FQ229469.1 | - | 5 - 6 | 848 | 94 | ||
FQ229972.1 | - | 5 - 6 | 849 | 94 | ||
FQ230190.1 | - | 5 - 6 | 848 | 94 | ||
FQ232129.1 | - | 5 - 6 | 861 | 94 | ||
FQ233579.1 | - | 5 - 6 | 848 | 94 | ||
FQ233966.1 | - | 5 - 6 | 848 | 94 | ||
FQ234183.1 | - | 5 - 6 | 850 | 94 | ||
FQ234500.1 | - | 5 - 6 | 848 | 94 | ||
FQ234529.1 | - | 5 - 6 | 848 | 94 | ||
FQ234547.1 | - | 5 - 6 | 848 | 94 | ||
FQ234986.1 | - | 5 - 6 | 848 | 94 | ||
FQ235296.1 | - | 5 - 6 | 848 | 94 | ||
Y00047.1 | - | 5 - 6 | 767 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Inventoried Probe spans exons |