Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpa3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111294.1 | NP_001104764.1 | 10 - 11 | 1237 | 79 | |
XM_008761911.1 | XP_008760133.1 | 10 - 11 | 1186 | 79 | ||
XM_008761913.1 | XP_008760135.1 | 10 - 11 | 1121 | 79 | ||
GenBank mRNA | CF110891.1 | - | 4 - 5 | 403 | 79 | |
Y16641.1 | - | 6 - 7 | 599 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Rattus norvegicus Probe spans exons |