Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fsip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013078.1 | NP_001013096.1 | 1 - 2 | 175 | 100 | |
XM_006234734.2 | XP_006234796.1 | 1 - 2 | 210 | 100 | ||
XM_006234735.2 | XP_006234797.1 | 1 - 2 | 210 | 100 | ||
GenBank mRNA | BC082080.1 | - | 1 - 2 | 175 | 7133293 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |