Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wipf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057192.2 | NP_476540.2 | 131 | 78 | ||
GenBank mRNA | BC072475.1 | - | 131 | 7100751 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |