Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scn9a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133289.1 | NP_579823.1 | 4 - 5 | 602 | 71 | |
GenBank mRNA | AF000368.1 | - | 4 - 5 | 602 | 71 | |
U79568.1 | - | 4 - 5 | 602 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Rattus norvegicus Probe spans exons |