Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nusap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107762.1 | NP_001101232.1 | 6 - 7 | 932 | 80 | |
XM_006234769.2 | XP_006234831.1 | 6 - 7 | 763 | 80 | ||
XM_008762102.1 | XP_008760324.1 | 5 - 6 | 673 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |