Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc3h15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001010963.1 | NP_001010963.1 | 1 - 2 | 123 | 132 | |
GenBank mRNA | AY377983.1 | - | 1 - 2 | 79 | 132 | |
BC088438.1 | - | 1 - 2 | 123 | 7126102 | 132 | |
FQ232115.1 | - | 1 - 2 | 202 | 132 | ||
FQ233565.1 | - | 1 - 2 | 225 | 132 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |