Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Depdc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029916.1 | NP_001025087.1 | 4 - 5 | 820 | 96 | |
XM_006234642.2 | XP_006234704.1 | 4 - 5 | 843 | 96 | ||
GenBank mRNA | BC097357.1 | - | 4 - 5 | 820 | 7442188 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |