Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108693.1 | NP_001102163.1 | 3 - 4 | 373 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |