Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tprg1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271085.1 | NP_001258014.1 | 2 - 3 | 621 | 106 | |
GenBank mRNA | BC105914.1 | - | 2 - 3 | 482 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons |