Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003750065.3 | XP_003750113.1 | 11 - 12 | 2503 | 64 | |
XM_006239290.2 | XP_006239352.1 | 11 - 12 | 2506 | 64 | ||
XM_006239291.2 | XP_006239353.1 | 11 - 12 | 2506 | 64 | ||
XM_006239292.2 | XP_006239354.1 | 11 - 12 | 2506 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |