Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003750065.3 | XP_003750113.1 | 13 - 14 | 2825 | 130 | |
XM_006239290.2 | XP_006239352.1 | 13 - 14 | 2831 | 130 | ||
XM_006239291.2 | XP_006239353.1 | 13 - 14 | 2836 | 130 | ||
XM_006239292.2 | XP_006239354.1 | 13 - 14 | 2833 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Rattus norvegicus Probe spans exons |