Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk11b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145766.2 | NP_665709.2 | 14 - 15 | 1617 | 81 | |
XM_006239527.2 | XP_006239589.1 | 13 - 14 | 2173 | 81 | ||
XM_006239528.2 | XP_006239590.1 | 14 - 15 | 1980 | 81 | ||
XM_006239529.2 | XP_006239591.1 | 14 - 15 | 1901 | 81 | ||
XM_006239530.1 | XP_006239592.1 | 14 - 15 | 1601 | 81 | ||
XM_006239531.2 | XP_006239593.1 | 14 - 15 | 1705 | 81 | ||
XM_006239532.2 | XP_006239594.1 | 9 - 10 | 2049 | 81 | ||
XM_006239533.1 | XP_006239595.1 | 9 - 10 | 1026 | 81 | ||
GenBank mRNA | DY544650.1 | - | 2 - 3 | 213 | 81 | |
EX490221.1 | - | 6 - 7 | 707 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Detect Genomic DNA Rattus norvegicus Probe spans exons |