Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pef1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007651.1 | NP_001007652.1 | 3 - 4 | 486 | 76 | |
GenBank mRNA | BC082028.1 | - | 3 - 4 | 486 | 7130077 | 76 |
FQ211313.1 | - | 3 - 4 | 483 | 76 | ||
FQ211450.1 | - | 3 - 4 | 499 | 76 | ||
FQ219120.1 | - | 3 - 4 | 499 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |