Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txlna | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006238972.2 | XP_006239034.1 | 1 - 2 | 122 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |