Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfyc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012866.2 | NP_036998.1 | 10 | 90 | ||
XM_006238785.2 | XP_006238847.1 | 10 | 90 | |||
GenBank mRNA | BC093619.1 | - | 10 | 7314492 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Both primers and probe map within a single exon |