Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002729537.3 | XP_002729583.1 | 5 - 6 | 836 | 86 | |
XM_006238790.2 | XP_006238852.1 | 5 - 6 | 839 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons |