Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053595.2 | NP_446047.1 | 4 - 5 | 687 | 76 | |
GenBank mRNA | BC087006.1 | - | 4 - 5 | 687 | 7193034 | 76 |
L40030.1 | - | 4 - 5 | 422 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |