Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc10a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC091152.1 | - | 2 - 3 | 636 | 7370832 | 72 |
FQ218559.1 | - | 2 - 3 | 648 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |