Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smek1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108050.2 | NP_001101520.2 | 14 - 15 | 2816 | 100 | |
NM_001271174.1 | NP_001258103.1 | 14 - 15 | 3113 | 100 | ||
GenBank mRNA | CO400159.1 | - | 2 - 3 | 256 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Rattus norvegicus 1700 Probe spans exons |