Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tgfb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013174.2 | NP_037306.1 | 2 - 3 | 912 | 66 | |
XM_006240366.1 | XP_006240428.1 | 3 - 4 | 1256 | 66 | ||
GenBank mRNA | BC092195.1 | - | 2 - 3 | 899 | 7313608 | 66 |
U03491.1 | - | 2 - 3 | 912 | 66 | ||
X71903.1 | - | 1 - 2 | 40 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |