Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Golga5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033065.1 | NP_001028237.1 | 7 - 8 | 1605 | 73 | |
XM_006240461.2 | XP_006240523.1 | 8 - 9 | 1860 | 73 | ||
GenBank mRNA | AY144587.1 | - | 7 - 8 | 1605 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |