Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpina11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166352.1 | NP_001159824.1 | 1 - 2 | 103 | 98 | |
XM_006240481.2 | XP_006240543.1 | 1 - 2 | 102 | 98 | ||
GenBank mRNA | EX492846.1 | - | 1 - 2 | 103 | 98 | |
FQ211426.1 | - | 1 - 2 | 75 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |