Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptgr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015009.1 | NP_001015009.1 | 3 - 4 | 324 | 62 | |
XM_006240326.1 | XP_006240388.1 | 3 - 4 | 338 | 62 | ||
XM_006240327.1 | XP_006240389.1 | 3 - 4 | 274 | 62 | ||
XM_006240328.2 | XP_006240390.1 | 3 - 4 | 236 | 62 | ||
GenBank mRNA | BC091173.1 | - | 3 - 4 | 324 | 7189599 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |