Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ift43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134525.1 | NP_001127997.1 | 7 - 8 | 492 | 78 | |
XM_006240370.2 | XP_006240432.1 | 8 - 9 | 562 | 78 | ||
XM_006240372.2 | XP_006240434.1 | 7 - 8 | 478 | 78 | ||
GenBank mRNA | FQ222650.1 | - | 3 - 4 | 213 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |