Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gar1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024306.1 | NP_001019477.1 | 4 - 5 | 707 | 144 | |
XM_006232006.2 | XP_006232068.1 | 4 - 5 | 1435 | 144 | ||
XM_008761519.1 | XP_008759741.1 | 4 - 5 | 1436 | 144 | ||
GenBank mRNA | BC079131.1 | - | 4 - 5 | 707 | 7127379 | 144 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |